Anti-polA Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46468-100UG A46468-50UG
77711-253EA 529.22 CAD
77711-253 77711-254
Anti-polA Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: polA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Thermus aquaticus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR