Anti-PNOC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47104-50UL A47104-100UL
77712-382EA 360.84 CAD
77712-382 77712-381
Anti-PNOC Rabbit Polyclonal Antibody
ELISA:1:2000-1:10000, WB:1:1000-1:5000

Type:
Antigen: PNOC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR