Anti-PAFAH2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46781-100UL A46781-50UL
77711-819EA 529.22 CAD
77711-819 77711-820
Anti-PAFAH2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:200-1:1000, IHC:1:100-1:300

Type:
Antigen: PAFAH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR