Anti-NTH1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47008-50UG A47008-100UG
77712-216EA 360.84 CAD
77712-216 77712-215
Anti-NTH1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: NTH1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR