Anti-NFKBID Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46708-100UL A46708-50UL
77711-682EA 529.22 CAD
77711-682 77711-683
Anti-NFKBID Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: NFKBID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR