Anti-Muc5ac Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46648-50UG A46648-100UG
77711-575EA 360.84 CAD
77711-575 77711-574
Anti-Muc5ac Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Muc5ac
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR