Anti-MT4 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46754-50UG A46754-100UG
77711-770EA 360.84 CAD
77711-770 77711-769
Anti-MT4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: MT4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR