Anti-MED22 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47179-50UL A47179-100UL
77712-511EA 360.84 CAD
77712-511 77712-510
Anti-MED22 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: MED22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR