Anti-MAGEA4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46939-50UL A46939-100UL
77712-092EA 360.84 CAD
77712-092 77712-091
Anti-MAGEA4 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:500-1:2000, IHC:1:5-1:20

Type:
Antigen: MAGEA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR