Anti-LOX Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46555-100UG A46555-50UG
77711-403EA 529.22 CAD
77711-403 77711-404
Anti-LOX Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: LOX
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR