Anti-LCP1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46768-100UL A46768-50UL
77711-795EA 529.22 CAD
77711-795 77711-796
Anti-LCP1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000

Type:
Antigen: LCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR