Anti-KCNK9 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46784-100UL A46784-50UL
77711-825EA 529.22 CAD
77711-825 77711-826
Anti-KCNK9 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:100-1:300

Type:
Antigen: KCNK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR