Anti-KAT8 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46860-50UG A46860-100UG
77711-950EA 360.84 CAD
77711-950 77711-949
Anti-KAT8 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: KAT8
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR