Anti-KARS Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46358-50UL A46358-100UL
77711-060EA 360.84 CAD
77711-060 77711-059
Anti-KARS Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:100-1:300

Type:
Antigen: KARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR