Anti-IL18BP Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46490-50UG A46490-100UG
77711-289EA 360.84 CAD
77711-289 77711-288
Anti-IL18BP Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: IL18BP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR