Anti-IL12A Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A46524-50UG A46524-100UG
77711-347EA 360.84 CAD
77711-347 77711-346
Anti-IL12A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: IL12A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR