Anti-HRASLS2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46217-100UL A46217-50UL
77710-807EA 529.22 CAD
77710-807 77710-808
Anti-HRASLS2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:200-1:1000, IHC:1:100-1:300

Type:
Antigen: HRASLS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR