Anti-Hp Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46611-50UG A46611-100UG
77711-505EA 360.84 CAD
77711-505 77711-504
Anti-Hp Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Hp
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR