Anti-HLA-DRB1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47268-100UG A47268-50UG
77689-248EA 529.22 CAD
77689-248 77712-661
Anti-HLA-DRB1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200

Type:
Antigen: HLA-DRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR