Anti-HDAC9 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46893-100UG A46893-50UG
77712-007EA 529.22 CAD
77712-007 77712-008
Anti-HDAC9 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: HDAC9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR