Anti-Havcr1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46619-100UG A46619-50UG
77711-520EA 529.22 CAD
77711-520 77711-521
Anti-Havcr1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Havcr1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR