Anti-GNG2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46390-50UL A46390-100UL
77711-115EA 360.84 CAD
77711-115 77711-114
Anti-GNG2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200

Type:
Antigen: GNG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR