Anti-glpK Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47092-50UG A47092-100UG
77712-361EA 360.84 CAD
77712-361 77712-360
Anti-glpK Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: glpK
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR