Anti-FOXA2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46501-50UL A46501-100UL
77711-306EA 360.84 CAD
77711-306 77711-305
Anti-FOXA2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000

Type:
Antigen: FOXA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR