Anti-fbpA Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47051-50UG A47051-100UG
77712-289EA 360.84 CAD
77712-289 77712-288
Anti-fbpA Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: fbpA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mycobacterium tuberculosis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR