Anti-eutC Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47036-100UG A47036-50UG
77712-266EA 529.22 CAD
77712-266 77712-267
Anti-eutC Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: eutC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR