Anti-EREG Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47209-100UG A47209-50UG
77705-342EA 400.93 CAD
77705-342 77712-564
Anti-EREG Rabbit Polyclonal Antibody
Antibodies
IHC:1:50-300, ELISA:1:10000-20000

Type:
Antigen: EREG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR