Anti-DEGS1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47267-100UL A47267-50UL
77712-659EA 529.22 CAD
77712-659 77712-660
Anti-DEGS1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: DEGS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR