Anti-COL17A1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46976-50UG A46976-100UG
77712-154EA 360.84 CAD
77712-154 77712-153
Anti-COL17A1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: COL17A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR