Anti-CNGA3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47131-50UL A47131-100UL
77712-428EA 360.84 CAD
77712-428 77712-427
Anti-CNGA3 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000

Type:
Antigen: CNGA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR