Anti-CCL17 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46748-100UL A46748-50UL
77711-757EA 529.22 CAD
77711-757 77711-758
Anti-CCL17 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200

Type:
Antigen: CCL17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR