Anti-CASP8 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46984-50UG A46984-100UG
77712-169EA 360.84 CAD
77712-169 77712-168
Anti-CASP8 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: CASP8
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR