Anti-CALCA Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A47059-50UL A47059-100UL
77712-304EA 360.84 CAD
77712-304 77712-303
Anti-CALCA Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR