Anti-CACNB2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47257-50UL A47257-100UL
77712-642EA 360.84 CAD
77712-642 77712-641
Anti-CACNB2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: CACNB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR