Anti-ATRN Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47115-50UL A47115-100UL
77712-399EA 360.84 CAD
77712-399 77712-398
Anti-ATRN Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: ATRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR