Anti-ATP1A1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46994-50UG A46994-100UG
77712-189EA 360.84 CAD
77712-189 77712-188
Anti-ATP1A1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: ATP1A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR