Anti-ARHGDIA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47281-50UL A47281-100UL
77712-686EA 360.84 CAD
77712-686 77712-685
Anti-ARHGDIA Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:100-1:300

Type:
Antigen: ARHGDIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR