Anti-AQP2 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46274-50UG A46274-100UG
77710-905EA 360.84 CAD
77710-905 77710-904
Anti-AQP2 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: AQP2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR