Anti-AKT2 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46216-100UG A46216-50UG
77710-805EA 529.22 CAD
77710-805 77710-806
Anti-AKT2 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: AKT2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR