Anti-AKR1C1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47202-100UL A47202-50UL
77712-552EA 529.22 CAD
77712-552 77712-553
Anti-AKR1C1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200

Type:
Antigen: AKR1C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR