Anti-STX12 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60530-50UL A60530-100UL
77716-692EA 360.84 CAD
77716-692 77716-691
Anti-STX12 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: STX12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR