Anti-MON1A Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A60507-100UG A60507-50UG
77716-648EA 529.22 CAD
77716-648 77716-649
Anti-MON1A Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: MON1A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR