Anti-MON1A Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A60506-100UG A60506-50UG
77716-646EA 529.22 CAD
77716-646 77716-647
Anti-MON1A Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: MON1A
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR