Anti-MON1A Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A60505-100UG A60505-50UG
77716-644EA 529.22 CAD
77716-644 77716-645
Anti-MON1A Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: MON1A
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR