Anti-MON1A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60504-100UG A60504-50UG
77690-015EA 529.22 CAD
77690-015 77716-643
Anti-MON1A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200

Type:
Antigen: MON1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR