Anti-SERPINA11 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60480-100UG A60480-50UG
77690-014EA 529.22 CAD
77690-014 77716-596
Anti-SERPINA11 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: SERPINA11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR