Anti-VMO1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60443-100UG A60443-50UG
77690-010EA 529.22 CAD
77690-010 77716-526
Anti-VMO1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: VMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR