Anti-FHL2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60404-100UL A60404-50UL
77716-459EA 529.22 CAD
77716-459 77716-460
Anti-FHL2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:100-1:300

Type:
Antigen: FHL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR