Anti-USP9X Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60302-100UL A60302-50UL
77716-289EA 529.22 CAD
77716-289 77716-290
Anti-USP9X Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: USP9X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR