Anti-STYK1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60278-100UL A60278-50UL
77716-245EA 529.22 CAD
77716-245 77716-246
Anti-STYK1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000

Type:
Antigen: STYK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR